8-Bromoimidazo[1,2-a]pyridine-6-carbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 8-Bromo-6-cyanoimidazo[1,2-a]pyridine

054530-500MG 054530-1G 054530-5G
101907-650EA 556.92 USD
101907-650 101907-652 101907-654
8-Bromoimidazo[1,2-a]pyridine-6-carbonitrile ≥95%
8-Bromoimidazo[1,2-a]pyridine-6-carbonitrile

Hazard: Irritant

Formula: C₈H₄BrN₃
MW: 222.05 g/mol
Melting Pt: 182…184 °C
MDL Number: MFCD14584821
CAS Number: 1221791-93-0

Order Now

Specification Test Results

Melting point 182-184°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR