3-Cyano-2,7-dimethylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid ≥95%

Supplier: Matrix Scientific

054527-500MG 054527-1G
101907-638EA 747.69 USD
101907-638 101907-640
3-Cyano-2,7-dimethylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid ≥95%
3-Cyano-2,7-dimethylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₈N₄O₂
Melting Pt: 295 °C (dec.)
MDL Number: MFCD14584818
CAS Number: 1221792-41-1

Order Now

Specification Test Results

Melting point 295°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR