1-Thia-4-azaspiro[4.6]undecan-3-one ≥95%

Supplier: Matrix Scientific

054526-500MG 054526-1G
101907-634EA 278.46 USD
101907-634 101907-636
1-Thia-4-azaspiro[4.6]undecan-3-one ≥95%
1-Thia-4-azaspiro[4.6]undecan-3-one

Hazard: Irritant

Formula: C₉H₁₅NOS
Melting Pt: 151…153 °C
MDL Number: MFCD00101409
CAS Number: 1221791-73-6

Order Now

Specification Test Results

Melting point 151-153°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR