3-Chloro-5-cyano-2-pyridine carboxylic acid ≥95%

Supplier: Matrix Scientific

054525-500MG 054525-1G
101907-630EA 804.62 USD
101907-630 101907-632
3-Chloro-5-cyano-2-pyridine carboxylic acid ≥95%
3-Chloro-5-cyano-2-pyridine carboxylic acid

Hazard: Irritant

Formula: C₇H₃ClN₂O₂
MW: 182.57 g/mol
Melting Pt: 144 °C (dec.)
MDL Number: MFCD14584817
CAS Number: 1200497-81-9

Order Now

Specification Test Results

Melting point 144°CC(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR