8-Chloroimidazo[1,2-a]pyridine-6-carbonitrile ≥95%

Supplier: Matrix Scientific

054524-500MG 054524-1G 054524-5G
101907-624EA 423.08 USD
101907-624 101907-626 101907-628
8-Chloroimidazo[1,2-a]pyridine-6-carbonitrile ≥95%
8-Chloroimidazo[1,2-a]pyridine-6-carbonitrile

Hazard: Irritant

Formula: C₈H₄ClN₃
Melting Pt: 162…164 °C
MDL Number: MFCD14584816
CAS Number: 1221792-07-9

Order Now

Specification Test Results

Melting point 162-164°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR