1-Thia-4-azaspiro[4.5]decan-3-one ≥95%

Supplier: Matrix Scientific

054521-500MG 054521-1G
101907-612EA 278.46 USD
101907-612 101907-614
1-Thia-4-azaspiro[4.5]decan-3-one ≥95%
1-Thia-4-azaspiro[4.5]decan-3-one

Hazard: Irritant

Formula: C₈H₁₃NOS
Melting Pt: 166…168 °C
MDL Number: MFCD00101403
CAS Number: 4580-63-6

Order Now

Specification Test Results

Melting point 166-168°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR