3-Cyano-6-phenyl-2-pyridine carboxylic acid ≥95%

Supplier: Matrix Scientific

054515-500MG 054515-1G
101907-588EA 921.54 USD
101907-588 101907-590
3-Cyano-6-phenyl-2-pyridine carboxylic acid ≥95%
3-Cyano-6-phenyl-2-pyridine carboxylic acid

Hazard: Irritant

Formula: C₁₃H₈N₂O₂
MW: 224.22 g/mol
Melting Pt: 173 °C (dec.)
MDL Number: MFCD14584810
CAS Number: 1221792-38-6

Order Now

Specification Test Results

Melting point 173°CC(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR