Methyl-2-piperazino-4-(trifluoromethyl)nicotinate ≥95%

Supplier: Matrix Scientific

054513-500MG 054513-1G
101907-580EA 921.54 USD
101907-580 101907-582
Methyl-2-piperazino-4-(trifluoromethyl)nicotinate ≥95%
Methyl-2-piperazino-4-(trifluoromethyl)nicotinate

Hazard: Irritant

Formula: C₁₂H₁₄F₃N₃O₂
Melting Pt: 62…63 °C
MDL Number: MFCD14584808
CAS Number: 1221791-85-0

Order Now

Specification Test Results

Melting point 62-63°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR