2-Amino-3-chloro-5-(trifluoromethyl)isonicotinonitrile ≥95%

Supplier: Matrix Scientific

054505-500MG 054505-1G
101907-550EA 600 USD
101907-550 101907-552
2-Amino-3-chloro-5-(trifluoromethyl)isonicotinonitrile ≥95%
2-Amino-3-chloro-5-(trifluoromethyl)isonicotinonitrile

Hazard: Irritant

Formula: C₇H₃ClF₃N₃
MW: 221.57 g/mol
Melting Pt: 210…211 °C
MDL Number: MFCD14584801
CAS Number: 1221572-26-4

Order Now

Specification Test Results

Melting point 210-211°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR