2,3,5-Trichloroisonicotinonitrile ≥95%
Supplier: Matrix Scientific
054504-500MG
054504-1G
101907-546EA
921.54
USD
101907-546
101907-548
2,3,5-Trichloroisonicotinonitrile ≥95%
2,3,5-Trichloroisonicotinonitrile 2-(4-Formylphenyl)isonicotinonitrile
Hazard: Irritant
Formula:
C₆HCl₃N₂ Melting Pt: 58-59°C |
MDL Number:
MFCD14584800 CAS Number: 1221791-83-8 |
Specification Test Results
Melting point | 58-59°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...