3-Chloro-2-methoxy-5-(trifluoromethyl)isonicotinic acid ≥95%

Supplier: Matrix Scientific

054503-500MG 054503-1G
101907-542EA 600 USD
101907-542 101907-544
3-Chloro-2-methoxy-5-(trifluoromethyl)isonicotinic acid ≥95%
3-Chloro-2-methoxy-5-(trifluoromethyl)isonicotinic acid

Hazard: Irritant

Formula: C₈H₅ClF₃NO₃
Melting Pt: 143…144 °C
MDL Number: MFCD14584799
CAS Number: 1221792-35-3

Order Now

Specification Test Results

Melting point 143-144°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR