Methyl-7-methoxy-2-methyl-1,3-benzoxazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

054500-500MG 054500-1G
101907-530EA 556.92 USD
101907-530 101907-532
Methyl-7-methoxy-2-methyl-1,3-benzoxazole-5-carboxylate ≥95%
Methyl-7-methoxy-2-methyl-1,3-benzoxazole-5-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₁NO₄
MW: 221.21 g/mol
Melting Pt: 134…136 °C
MDL Number: MFCD14584796
CAS Number: 1197944-26-5

Order Now

Specification Test Results

Melting point 134-136°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR