Methyl-7-methoxy-1,3-benzoxazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

054497-500MG 054497-1G
101907-516EA 556.92 USD
101907-516 101907-518
Methyl-7-methoxy-1,3-benzoxazole-5-carboxylate ≥95%
Methyl-7-methoxy-1,3-benzoxazole-5-carboxylate

Hazard: Irritant

Formula: C₁₀H₉NO₄
Melting Pt: 135…136 °C
MDL Number: MFCD14584793
CAS Number: 1221792-20-6

Order Now

Specification Test Results

Melting point 135-136°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR