2-Bromo-4,6-diiodo-3-methoxypyridine ≥95%

Supplier: Matrix Scientific

054495-500MG 054495-1G
101907-508EA 375.38 USD
101907-508 101907-510
2-Bromo-4,6-diiodo-3-methoxypyridine ≥95%
2-Bromo-4,6-diiodo-3-methoxypyridine

Hazard: Irritant

Formula: C₆H₄BrI₂NO
Melting Pt: 124…126 °C
MDL Number: MFCD14584791
CAS Number: 1221792-52-4

Order Now

Specification Test Results

Melting point 124-126°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR