Benzyl-6-(bromomethyl)-2-chloro-3-pyridinyl ether ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(Benzyloxy)-6-(bromomethyl)-2-chloropyridine

054493-500MG 054493-1G
101907-500EA 747.69 USD
101907-500 101907-502
Benzyl-6-(bromomethyl)-2-chloro-3-pyridinyl ether ≥95%
Benzyl-6-(bromomethyl)-2-chloro-3-pyridinyl ether

Hazard: Irritant

Formula: C₁₃H₁₁BrClNO
MW: 312.6 g/mol
Melting Pt: 130…132 °C
MDL Number: MFCD14584789
CAS Number: 1221791-60-1

Order Now

Specification Test Results

Melting point 130-132°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR