Methyl-3-amino-4-(benzyloxy)-5-methoxybenzenecarboxylate ≥95%
Supplier: Matrix Scientific
054491-500MG
054491-1G
054491-5G
101907-490EA
556.92
USD
101907-490
101907-492
101907-494
Methyl-3-amino-4-(benzyloxy)-5-methoxybenzenecarboxylate ≥95%
Methyl-3-amino-4-(benzyloxy)-5-methoxybenzenecarboxylate
Hazard: Irritant
Formula:
C₁₆H₁₇NO₄ Melting Pt: 89…91 °C |
MDL Number:
MFCD14584787 CAS Number: 1221791-79-2 |
Specification Test Results
Melting point | 89-91°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...