5-Chloro-2-hydroxy-3-nitrophenylbenzene carboxylate ≥95%

Supplier: Matrix Scientific

054488-500MG 054488-1G
101907-480EA 556.92 USD
101907-480 101907-482
5-Chloro-2-hydroxy-3-nitrophenylbenzene carboxylate ≥95%
5-Chloro-2-hydroxy-3-nitrophenylbenzene carboxylate

Hazard: Irritant

Formula: C₁₃H₈ClNO₅
MW: 293.67 g/mol
Melting Pt: 95…96 °C
MDL Number: MFCD14584784
CAS Number: 194294-25-2

Order Now

Specification Test Results

Melting point 95-96°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR