Methyl-4-(benzyloxy)-3-methoxy-5-nitrobenzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-(benzyloxy)-3-methoxy-5-nitrobenzoate

054486-500MG 054486-1G 054486-5G
101907-468EA 375.38 USD
101907-468 101907-470 101907-472
Methyl-4-(benzyloxy)-3-methoxy-5-nitrobenzenecarboxylate ≥95%
Methyl-4-(benzyloxy)-3-methoxy-5-nitrobenzenecarboxylate

Hazard: Irritant

Formula: C₁₆H₁₅NO₆
MW: 317.3 g/mol
Melting Pt: 96…98 °C
MDL Number: MFCD14584782
CAS Number: 1221791-58-7

Order Now

Specification Test Results

Melting point 96-98°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR