Methyl-3-amino-4-hydroxy-5-methoxybenzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-amino-4-hydroxy-5-methoxybenzoate

054485-500MG 054485-1G 054485-5G
101907-462EA 261.54 USD
101907-462 101907-464 101907-466
Methyl-3-amino-4-hydroxy-5-methoxybenzenecarboxylate ≥95%
Methyl-3-amino-4-hydroxy-5-methoxybenzenecarboxylate

Hazard: Irritant

Formula: C₉H₁₁NO₄
MW: 197.19 g/mol
Melting Pt: 129…131 °C
MDL Number: MFCD12913545
CAS Number: 92643-72-6

Order Now

Specification Test Results

Melting point 129-131°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR