2-Bromo-3,6-dihydroxybenzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

054483-500MG 054483-1G 054483-5G
101907-450EA 170.77 USD
101907-450 101907-452 101907-454
2-Bromo-3,6-dihydroxybenzenecarbaldehyde ≥95%
2-Bromo-3,6-dihydroxybenzenecarbaldehyde

Hazard: Irritant

Formula: C₇H₅BrO₃
MW: 217.03 g/mol
Melting Pt: 137…138 °C
MDL Number: MFCD14584781
CAS Number: 241127-72-0

Order Now

Specification Test Results

Melting point 137-138°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR