4-Bromo-2-methyl-3-quinolinol ≥95%

Supplier: Matrix Scientific

054482-500MG 054482-1G
101907-446EA 278.46 USD
101907-446 101907-448
4-Bromo-2-methyl-3-quinolinol ≥95%
4-Bromo-2-methyl-3-quinolinol

Hazard: Irritant

Formula: C₁₀H₈BrNO
MW: 238.08 g/mol
Melting Pt: 164…166 °C
CAS Number: 13235-12-6

Order Now

Specification Test Results

Melting point 164-166°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR