3-Nitro-1,2-benzenediol ≥95%
Supplier: Matrix Scientific
054480-1G
054480-5G
101907-442EA
144.62
USD
101907-442
102833-190
3-Nitro-1,2-benzenediol ≥95%
3-Nitro-1,2-benzenediol
Hazard: Irritant
Formula:
C₆H₅NO₄ Melting Pt: 85…86 °C |
MDL Number:
MFCD00463758 CAS Number: 6665-98-1 |
Specification Test Results
Melting point | 85-86°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...