2-Hydroxyphenyl benzenecarboxylate ≥95%

Supplier: Matrix Scientific

054478-500MG 054478-1G 054478-5G
101907-428EA 181.54 USD
101907-428 101907-430 101907-432
2-Hydroxyphenyl benzenecarboxylate ≥95%
2-Hydroxyphenyl benzenecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₀O₃
MW: 214.22 g/mol
Melting Pt: 114…116 °C
MDL Number: MFCD00089689
CAS Number: 5876-92-6

Order Now

Specification Test Results

Melting point 114-116°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR