4,5-Difluoro-o-toluidine ≥98%

Supplier: Matrix Scientific

Synonyms: 4,5-Difluoro-2-methylaniline

054385-5G 054385-25G
101907-230EA 90.77 USD
101907-230 101907-232
4,5-Difluoro-o-toluidine ≥98%
4,5-Difluoro-o-toluidine

Hazard: Irritant

Formula: C₇H₇F₂N
MW: 143.14 g/mol
Melting Pt: 56-58°C
MDL Number: MFCD08062378
CAS Number: 875664-57-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 56-58°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR