2-[(3-Chlorobenzyl)sulfanyl]ethylamine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 2-[(3-Chlorobenzyl)thio]ethylamine hydrochloride

047730-500MG 047730-1G
101881-140EA 181.54 USD
101881-140 101881-142
2-[(3-Chlorobenzyl)sulfanyl]ethylamine hydrochloride ≥95%
2-[(3-Chlorobenzyl)sulfanyl]ethylamine hydrochloride

Hazard: Irritant

Formula: C₉H₁₃Cl₂NS
MW: 238.18 g/mol
MDL Number: MFCD09027132
CAS Number: 1170562-27-2

Order Now

Specification Test Results

Melting point 118-120°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR