5-Nitro-2-(3-thienyl)pyridine ≥95%

Supplier: Matrix Scientific

047729-500MG 047729-1G 047729-5G
101881-134EA 278.46 USD
101881-134 101881-136 101881-138
5-Nitro-2-(3-thienyl)pyridine ≥95%
5-Nitro-2-(3-thienyl)pyridine

Hazard: Irritant

Formula: C₉H₆N₂O₂S
MW: 206.22 g/mol
Melting Pt: 155…157 °C
MDL Number: MFCD09027131
CAS Number: 937602-39-6

Order Now

Specification Test Results

Melting point 155-157°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR