2-(3-Bromo-2-thienyl)-2-oxoethyl thiocyanate ≥95%

Supplier: Matrix Scientific

047728-500MG 047728-1G 047728-5G
101881-128EA 556.92 USD
101881-128 101881-130 101881-132
2-(3-Bromo-2-thienyl)-2-oxoethyl thiocyanate ≥95%
2-(3-Bromo-2-thienyl)-2-oxoethyl thiocyanate

Hazard: Irritant

Formula: C₇H₄BrNOS₂
MW: 262.15 g/mol
Melting Pt: 99…100 °C
MDL Number: MFCD09027130
CAS Number: 937602-37-4

Order Now

Specification Test Results

Melting point 99-100°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR