2-(3-Chloro-2-thienyl)-2-oxoethyl thiocyanate ≥95%

Supplier: Matrix Scientific

047727-500MG 047727-1G 047727-5G
101881-122EA 921.54 USD
101881-122 101881-124 101881-126
2-(3-Chloro-2-thienyl)-2-oxoethyl thiocyanate ≥95%
2-(3-Chloro-2-thienyl)-2-oxoethyl thiocyanate

Hazard: Irritant

Formula: C₇H₄ClNOS₂
MW: 217.7 g/mol
Melting Pt: 76…78 °C
MDL Number: MFCD09027129
CAS Number: 937602-35-2

Order Now

Specification Test Results

Melting point 76-78°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR