2,4-Dichlorophenacyl thiocyanate ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(2,4-Dichlorophenyl)-2-oxoethyl thiocyanate
047724-500MG
047724-1G
047724-5G
101881-104EA
181.54
USD
101881-104
101881-106
101881-108
2,4-Dichlorophenacyl thiocyanate ≥95%
2,4-Dichlorophenacyl thiocyanate
Hazard: Irritant
Formula:
C₉H₅Cl₂NOS Melting Pt: 100…102 °C |
MDL Number:
MFCD03453108 CAS Number: 125488-14-4 |
Specification Test Results
Melting point | 100-102°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...