2-(4-Fluorophenyl)-2-oxoethyl thiocyanate ≥95%

Supplier: Matrix Scientific

047722-500MG 047722-1G 047722-5G
101881-092EA 181.54 USD
101881-092 101881-094 101881-096
2-(4-Fluorophenyl)-2-oxoethyl thiocyanate ≥95%
2-(4-Fluorophenyl)-2-oxoethyl thiocyanate

Hazard: Irritant

Formula: C₉H₆FNOS
MW: 195.22 g/mol
Melting Pt: 99…101 °C
MDL Number: MFCD02029655
CAS Number: 43045-16-5

Order Now

Specification Test Results

Melting point 99-101°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR