4-Methylphenacyl thiocyanate ≥97%
Supplier: Matrix Scientific
Synonyms:
2-(4-Methylphenyl)-2-oxoethyl thiocyanate
047720-500MG
047720-1G
047720-5G
101881-082EA
135.38
USD
101881-082
101881-084
101881-086
4-Methylphenacyl thiocyanate ≥97%
4-Methylphenacyl thiocyanate
Hazard: Irritant
Formula:
C₁₀H₉NOS Melting Pt: 105…107 °C |
MDL Number:
MFCD00278207 CAS Number: 6097-27-4 |
Specification Test Results
Melting point | 105-107°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...