4-[3-(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)propoxy]-N'-hydroxybenzenecarboximidamide ≥95%

Supplier: Matrix Scientific

047717-500MG 047717-1G 047717-5G
101881-064EA 278.46 USD
101881-064 101881-066 101881-068
4-[3-(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)propoxy]-N'-hydroxybenzenecarboximidamide ≥95%
4-[3-(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)propoxy]-N'-hydroxybenzenecarboximidamide

Hazard: Irritant

Formula: C₁₈H₁₇N₃O₄
Melting Pt: 197…199 °C
MDL Number: MFCD09027124
CAS Number: 145259-45-6

Order Now

Specification Test Results

Melting point 197-199°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR