3-Benzyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%

Supplier: Matrix Scientific

047710-500MG 047710-1G
101881-040EA 90.77 USD
101881-040 101881-042
3-Benzyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%
3-Benzyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone

Hazard: Irritant

Formula: C₁₅H₁₂N₂OS
Melting Pt: >250 °C
MDL Number: MFCD00125914
CAS Number: 13906-05-3

Order Now

Specification Test Results

Melting point >250°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR