3-(4-Fluorophenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%

Supplier: Matrix Scientific

047706-500MG 047706-1G
101881-030EA 181.54 USD
101881-030 101881-032
3-(4-Fluorophenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%
3-(4-Fluorophenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone

Hazard: Irritant

Formula: C₁₄H₉FN₂OS
MW: 272.3 g/mol
Melting Pt: >250 °C
MDL Number: MFCD00807054
CAS Number: 1547-15-5

Order Now

Specification Test Results

Melting point >250°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR