Methyl-2-(2,5-dioxo-1-pyrrolidinyl)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-(2,5-dioxopyrrolidin-1-yl)benzoate

047704-500MG 047704-1G
101881-024EA 181.54 USD
101881-024 101881-026
Methyl-2-(2,5-dioxo-1-pyrrolidinyl)benzenecarboxylate ≥95%
Methyl-2-(2,5-dioxo-1-pyrrolidinyl)benzenecarboxylate

Hazard: Irritant

Formula: C₁₂H₁₁NO₄
MW: 233.23 g/mol
Melting Pt: 79…80 °C
MDL Number: MFCD01158843
CAS Number: 77741-53-8

Order Now

Specification Test Results

Melting point 79-80°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR