4-[(4-Fluorophenyl)sulfanyl]-3-nitrobenzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

047702-500MG 047702-1G
101881-020EA 181.54 USD
101881-020 101881-022
4-[(4-Fluorophenyl)sulfanyl]-3-nitrobenzenecarbaldehyde ≥95%
4-[(4-Fluorophenyl)sulfanyl]-3-nitrobenzenecarbaldehyde

Hazard: Irritant

Formula: C₁₃H₈FNO₃S
Melting Pt: 151…152 °C
MDL Number: MFCD09027121
CAS Number: 952182-59-1

Order Now

Specification Test Results

Melting point 151-152°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR