N'-(2-Cyano-4-nitrophenyl)-N,N-dimethyliminoformamide ≥95%

Supplier: Matrix Scientific

047701-500MG 047701-1G
101881-016EA 181.54 USD
101881-016 101881-018
N'-(2-Cyano-4-nitrophenyl)-N,N-dimethyliminoformamide ≥95%
N'-(2-Cyano-4-nitrophenyl)-N,N-dimethyliminoformamide

Hazard: Irritant

Formula: C₁₀H₁₀N₄O₂
Melting Pt: 146…148 °C
MDL Number: MFCD09027120
CAS Number: 39263-34-8

Order Now

Specification Test Results

Melting point 146-148°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR