2-Chloro-N-{4-[(2,4-dichlorobenzyl)sulfanyl]phenyl}acetamide ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-N-{4-[(2,4-dichlorobenzyl)thio]phenyl}acetamide

047699-500MG 047699-1G
101881-012EA 181.54 USD
101881-012 101881-014
2-Chloro-N-{4-[(2,4-dichlorobenzyl)sulfanyl]phenyl}acetamide ≥95%
2-Chloro-N-{4-[(2,4-dichlorobenzyl)sulfanyl]phenyl}acetamide

Hazard: Irritant

Formula: C₁₅H₁₂Cl₃NOS
MW: 360.69 g/mol
Melting Pt: 114…115 °C
MDL Number: MFCD09027119
CAS Number: 952182-58-0

Order Now

Specification Test Results

Melting point 114-115°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR