1-(4-Bromo-2-fluorophenyl)-1H-pyrrole-2,5-dione ≥95%

Supplier: Matrix Scientific

047695-500MG 047695-1G
101881-004EA 181.54 USD
101881-004 101881-006
1-(4-Bromo-2-fluorophenyl)-1H-pyrrole-2,5-dione ≥95%
1-(4-Bromo-2-fluorophenyl)-1H-pyrrole-2,5-dione

Hazard: Irritant

Formula: C₁₀H₅BrFNO₂
Melting Pt: 97…98 °C
MDL Number: MFCD08277325
CAS Number: 893614-85-2

Order Now

Specification Test Results

Melting point 97-98°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR