2-[(4-Chlorobenzyl)sulfanyl]-5-nitrobenzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(4-Chlorobenzylthio)-5-nitrobenzaldehyde

047680-500MG 047680-1G
101880-962EA 181.54 USD
101880-962 101880-964
2-[(4-Chlorobenzyl)sulfanyl]-5-nitrobenzenecarbaldehyde ≥95%
2-[(4-Chlorobenzyl)sulfanyl]-5-nitrobenzenecarbaldehyde

Hazard: Irritant

Formula: C₁₄H₁₀ClNO₃S
MW: 307.76 g/mol
Melting Pt: 154…155 °C
MDL Number: MFCD07021302
CAS Number: 319454-75-6

Order Now

Specification Test Results

Melting point 154-155°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR