4-(Cyclohexylsulfanyl)-3-nitrobenzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

047679-500MG 047679-1G
101880-958EA 181.54 USD
101880-958 101880-960
4-(Cyclohexylsulfanyl)-3-nitrobenzenecarbaldehyde ≥95%
4-(Cyclohexylsulfanyl)-3-nitrobenzenecarbaldehyde

Hazard: Irritant

Formula: C₁₃H₁₅NO₃S
Melting Pt: 98…99 °C
MDL Number: MFCD00098772
CAS Number: 270262-95-8

Order Now

Specification Test Results

Melting point 98-99°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR