2-[(2-Furylmethyl)sulfanyl]-5-nitrobenzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 2-[(Fur-2-ylmethyl)thio]-5-nitrobenzaldehyde

047676-500MG 047676-1G
101880-946EA 181.54 USD
101880-946 101880-948
2-[(2-Furylmethyl)sulfanyl]-5-nitrobenzenecarbaldehyde ≥95%
2-[(2-Furylmethyl)sulfanyl]-5-nitrobenzenecarbaldehyde

Hazard: Irritant

Formula: C₁₂H₉NO₄S
Melting Pt: 131.5…132.5 °C
MDL Number: MFCD07021300
CAS Number: 885267-52-7

Order Now

Specification Test Results

Melting point 131.5-132.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR