4-Isobutyl-5-phenyl-4H-1,2,4-triazol-3-ylhydrosulfide ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Isobutyl-5-phenyl-4H-1,2,4-triazole-3-thiol

047669-500MG 047669-1G
101880-922EA 181.54 USD
101880-922 101880-924
4-Isobutyl-5-phenyl-4H-1,2,4-triazol-3-ylhydrosulfide ≥95%
4-Isobutyl-5-phenyl-4H-1,2,4-triazol-3-ylhydrosulfide

Hazard: Irritant

Formula: C₁₂H₁₅N₃S
MW: 233.34 g/mol
Melting Pt: 187…189 °C
MDL Number: MFCD05134110
CAS Number: 26029-10-7

Order Now

Specification Test Results

Melting point 187-189°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR