1-(3-Chloro-4-fluorophenyl)biguanide hydrochloride ≥95%

Supplier: Matrix Scientific

047667-500MG 047667-1G
101880-916EA 181.54 USD
101880-916 101880-918
1-(3-Chloro-4-fluorophenyl)biguanide hydrochloride ≥95%
1-(3-Chloro-4-fluorophenyl)biguanide hydrochloride

Hazard: Irritant

Formula: C₈H₁₀Cl₂FN₅
Melting Pt: 89…89.5 °C
MDL Number: MFCD00126332
CAS Number: 915071-41-9

Order Now

Specification Test Results

Melting point 89-89.5°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR