1-(2-Methoxy-5-nitrophenyl)-1H-pyrrole-2,5-dione ≥95%

Supplier: Matrix Scientific

Synonyms: N-(2-Methoxy-5-nitrophenyl)maleimide

047654-500MG 047654-1G
101880-890EA 181.54 USD
101880-890 101880-892
1-(2-Methoxy-5-nitrophenyl)-1H-pyrrole-2,5-dione ≥95%
1-(2-Methoxy-5-nitrophenyl)-1H-pyrrole-2,5-dione

Hazard: Irritant

Formula: C₁₁H₈N₂O₅
Melting Pt: 158…159 °C
MDL Number: MFCD00523329
CAS Number: 17392-67-5

Order Now

Specification Test Results

Melting point 158-159°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR