Methyl-3,5-dibromo-2-methoxybenzenecarboxylate ≥95%

Supplier: Matrix Scientific

047646-500MG 047646-1G 047646-5G
101880-864EA 69.23 USD
101880-864 101880-866 101880-868
Methyl-3,5-dibromo-2-methoxybenzenecarboxylate ≥95%
Methyl-3,5-dibromo-2-methoxybenzenecarboxylate

Hazard: Irritant

Formula: C₉H₈Br₂O₃
Melting Pt: 53…54 °C
MDL Number: MFCD00189410
CAS Number: 15790-59-7

Order Now

Specification Test Results

Melting point 53-54°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR