1-(3-Methoxy-4-nitrophenyl)-1-ethanone ≥97%

Supplier: Matrix Scientific

Synonyms: 3'-Methoxy-4'-nitroacetophenone

047644-500MG 047644-1G 047644-5G
101880-854EA 375.38 USD
101880-854 101880-856 101880-858
1-(3-Methoxy-4-nitrophenyl)-1-ethanone ≥97%
1-(3-Methoxy-4-nitrophenyl)-1-ethanone

Hazard: Irritant

Formula: C₉H₉NO₄
MW: 195.18 g/mol
Melting Pt: 71…73 °C
MDL Number: MFCD00075788
CAS Number: 22106-39-4

Order Now

Specification Test Results

Melting point 71-73°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR