4'-(4-Fluorobenzyloxy)acetophenone ≥95%

Supplier: Matrix Scientific

Synonyms: 1-{4-[(4-Fluorobenzyl)oxy]phenyl}ethan-1-one

047640-500MG 047640-1G
101880-840EA 46.15 USD
101880-840 101880-842
4'-(4-Fluorobenzyloxy)acetophenone ≥95%
4'-(4-Fluorobenzyloxy)acetophenone

Hazard: Irritant

Formula: C₁₅H₁₃FO₂
Melting Pt: 78…79 °C
MDL Number: MFCD00173651
CAS Number: 72293-96-0

Order Now

Specification Test Results

Melting point 78-79°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR