2-(Benzylsulfanyl)-5-nitrobenzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Benzylthio)-5-nitrobenzaldehyde

047620-500MG 047620-1G 047620-5G
101880-806EA 181.54 USD
101880-806 101880-808 101880-810
2-(Benzylsulfanyl)-5-nitrobenzenecarbaldehyde ≥95%
2-(Benzylsulfanyl)-5-nitrobenzenecarbaldehyde

Hazard: Irritant

Formula: C₁₄H₁₁NO₃S
MW: 273.31 g/mol
Melting Pt: 141…143 °C
MDL Number: MFCD00173709
CAS Number: 175278-43-0

Order Now

Specification Test Results

Melting point 141-143°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR