1-(4-Fluorophenyl)-3-(4-hydroxyphenyl)-2-propen-1-one ≥95%

Supplier: Matrix Scientific

047614-500MG 047614-1G
101880-782EA 181.54 USD
101880-782 101880-784
1-(4-Fluorophenyl)-3-(4-hydroxyphenyl)-2-propen-1-one ≥95%
1-(4-Fluorophenyl)-3-(4-hydroxyphenyl)-2-propen-1-one

Hazard: Irritant

Formula: C₁₅H₁₁FO₂
MW: 242.25 g/mol
Melting Pt: 189…190 °C
MDL Number: MFCD00206942
CAS Number: 7397-22-0

Order Now

Specification Test Results

Melting point 189-190°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR